Antimony(III) bromide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-5112-250G 93-5112-50G
100203-502EA 751.57 USD
100203-502 100201-128
Antimony(III) bromide ≥99%
Antimony (III) bromide
Formula: Br₃Sb
Boiling Pt: 280 °C
Melting Pt: 96.6 °C
Density: 4.148
MDL Number: MFCD00016317
CAS Number: 7789-61-9
EINECS: 232-179-8
UN: 1549
ADR: 6.1,III
Merck Index: 12,00747

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR