Indium(III) trifluoroacetate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-4904-5G
100203-494EA 300.06 USD
100203-494
Indium(III) trifluoroacetate ≥99%
Indium(III) trifluoroacetate
MDL Number: MFCD00050940
CAS Number: 36554-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR