Cadmium ≥99.9%, pieces

Supplier: STREM CHEMICALS INC MS
Danger

93-4844-100G 93-4844-500G
100205-610EA 77.85 USD
100205-610 100203-492
Cadmium ≥99.9%, pieces
Cadmium
Formula: Cd
Boiling Pt: 765 °C
Melting Pt: 320.9 °C
Density: 8.642
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR