Rubidium bromide ≥99.9% (Rb basis)

Supplier: STREM CHEMICALS INC MS

93-3726-5G 93-3726-25G
100203-456EA 94.83 USD
100203-456 100210-690
Rubidium bromide ≥99.9% (Rb basis)
Rubidium bromide
Formula: BrRb
Boiling Pt: 1340 °C
Melting Pt: 682 °C
Density: 3.35
Storage Temperature: Ambient
MDL Number: MFCD00011186
CAS Number: 7789-39-1
EINECS: 232-162-5
Merck Index: 12,08440

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR