Selenium dioxide ≥99.8%

Supplier: STREM CHEMICALS INC MS
Danger

93-3406-500G 93-3406-100G
100203-454EA 646.83 USD
100203-454 100200-554
Selenium dioxide ≥99.8%
Selenium dioxide
Formula: SeO₂
Boiling Pt: 340…350 °C (subl.)
Melting Pt: 315 °C
Density: 3.95
Storage Temperature: Ambient
MDL Number: MFCD00003562
CAS Number: 7446-08-4
EINECS: 231-194-7
UN: 3283
ADR: 6.1,III
Merck Index: 13,08508

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR