Zinc chloride ≥99.99% (Zn basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

93-3052-50G 93-3052-10G
100203-448EA 781.29 USD
100203-448 100203-446
Zinc chloride ≥99.99% (Zn basis), PURATREM
Zinc chloride
Formula: ZnCl₂
Boiling Pt: 732 °C
Melting Pt: 283 °C
Density: 2.91
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR