Tetraiodosilane ≥99.9% (Si basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-1447-2G 93-1447-10G
100203-384EA 124.55 USD
100203-384 100208-110
Tetraiodosilane ≥99.9% (Si basis)
Tetraiodosilane
Formula: I₄Si
Boiling Pt: 287.5 °C
Melting Pt: 120.5 °C
Density: 4.198
MDL Number: MFCD00054041
CAS Number: 13465-84-4
EINECS: 236-706-2
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR