Sodium hexacyanoferrate(II) decahydrate
Supplier: STREM CHEMICALS INC MS
Synonyms:
Sodium ferrocyanide decahydrate
93-1140-500G
93-1140-2000G
100204-144EA
236.37
USD
100204-144
100203-360
Sodium hexacyanoferrate(II) decahydrate
Sodium hexacyanoferrate(II) decahydrate
Formula:
C₆H₂₀FeN₆Na₄O₁₀ Density: 1,458 |
MDL Number:
MFCD00085360 CAS Number: 14434-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...