Platinum ≥99.99%, wire

Supplier: STREM CHEMICALS INC MS

78-0075-100CM 78-0075-25CM
100208-058EA 530.77 USD
100208-058 100203-304
Platinum ≥99.99%, wire
Platinum
Formula: Pt
Boiling Pt: 3827 °C
Melting Pt: 1769 °C
Density: 21.45
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR