Ytterbium(III) isopropoxide ≥99.9% (Yb basis)

Supplier: STREM CHEMICALS INC MS
Danger

70-5000-5G 70-5000-1G
100206-784EA 529.35 USD
100206-784 100203-256
Ytterbium(III) isopropoxide ≥99.9% (Yb basis)
Ytterbium (III) isopropoxide

Alkoxide

Formula: C₉H₂₁O₃Yb
Boiling Pt: 190…200 °C (0,2 mmHg) (subl.)
Melting Pt: 200 °C (dec.)
MDL Number: MFCD00145532
CAS Number: 6742-69-4
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR