Silver(I) hexafluorophosphate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Silver hexafluorophosphate

47-1000-5G 47-1000-1G 47-1000-25G
100196-296EA 147.2 USD
100196-296 100197-550 100203-212
Silver(I) hexafluorophosphate ≥99%
Silver (I) hexafluorophosphate
Formula: AgF₆P
MDL Number: MFCD00003415
CAS Number: 26042-63-7
EINECS: 247-428-6
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR