Silver ≥99.9%, wire

Supplier: STREM CHEMICALS INC MS

47-0250-25MT 47-0250-1MT 47-0250-5MT
100203-210EA 1037.48 USD
100203-210 100203-208 100201-000
Silver ≥99.9%, wire
Silver
Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960.5 °C
Density: 10.49
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR