Ruthenium(III) chloride, anhydrous

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Ruthenium trichloride

44-5850-1G 44-5850-5G
100203-158EA 150.03 USD
100203-158 100199-916
Ruthenium(III) chloride, anhydrous
Ruthenium (III) chloride
Formula: Cl₃Ru
Melting Pt: >500 °C (dec.)
Density: 3.11 g/cm³ (20 °C)
MDL Number: MFCD00011208
CAS Number: 10049-08-8
EINECS: 233-167-5
UN: 1759
ADR: 8,II
Merck Index: 12,08455

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR