Yttrium(III) 2-ethylhexanoate ≥99.9% (Y basis) (15-17% Y), Superconductor Grade

Supplier: STREM CHEMICALS INC MS
Warning

39-2400-10G 39-2400-50G
100203-108EA 212.31 USD
100203-108 100203-110
Yttrium(III) 2-ethylhexanoate ≥99.9% (Y basis) (15-17% Y), Superconductor Grade
Yttrium (III) 2-ethylhexanoate
Formula: C₂₄H₄₅O₆Y
MDL Number: MFCD00070441
CAS Number: 103470-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR