Arsenic(III) oxide ≥99.95% ACS primary standard

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: di-Arsenic trioxide

33-3313-100G
100203-098EA 450.09 USD
100203-098
Arsenic(III) oxide ≥99.95% ACS primary standard
Arsenic (III) oxide
Formula: As₂O₃
Boiling Pt: 465 °C (subl.)
Melting Pt: 315 °C
Density: 3.738
Storage Temperature: Ambient
MDL Number: MFCD00003433
CAS Number: 1327-53-3
EINECS: 215-481-4
UN: 1561
ADR: 6.1,II
Merck Index: 12,00844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR