Zinc bromide ≥99.999% (Zn basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Zinc (II) bromide

30-3033-25G 30-3033-5G
100205-838EA 182.58 USD
100205-838 100203-088
Zinc bromide ≥99.999% (Zn basis), PURATREM
Zinc bromide
Formula: Br₂Zn
Boiling Pt: 650 °C
Melting Pt: 394 °C
Density: 4.201
Storage Temperature: Ambient
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 1759
ADR: 8,II
Merck Index: 13,10182

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR