Trioctylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: TOP, Tri-n-octylphosphine

15-6655-500G 15-6655-25G 15-6655-100G
100203-012EA 559.08 USD
100203-012 100198-674 100203-010
Trioctylphosphine ≥97%
Trioctylphosphine

Nanomaterials, Phosphine, Surfactants for Nano Synthesis

Formula: C₂₄H₅₁P
Boiling Pt: 175 °C (0.3 mmHg)
Melting Pt: 30 °C
Density: 0,83
Flash Pt: 280 °F
MDL Number: MFCD00015298
CAS Number: 4731-53-7
EINECS: 225-234-2
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR