Cyclohexylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger


Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC)

15-0952-50G
100202-970EA 680.8 USD
100202-970
Cyclohexylphosphine ≥97%
Cyclohexylphosphine

Phosphine

Boiling Pt: 145 °C
Density: 0,8750
MDL Number: MFCD00014297
CAS Number: 822-68-4
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR