Bis(3,5-dimethylphenyl)phosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

15-0088-500MG
100202-936EA 292.98 USD
100202-936
Bis(3,5-dimethylphenyl)phosphine ≥98%
Bis(3,5-dimethylphenyl)phosphine
MDL Number: MFCD01074629
CAS Number: 71360-06-0
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR