2-(Trifluoromethyl)propene ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

09-7760-10G
100202-912EA 1307.82 USD
100202-912
2-(Trifluoromethyl)propene ≥97%
2-(Trifluoromethyl)propene

Organo Fluorine

Formula: C₄H₅F₃
Boiling Pt: 6…7 °C
MDL Number: MFCD00042093
CAS Number: 374-00-5
UN: 1954
ADR: 2.1,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR