1,3-Bis(1-adamantyl)imidazolium chloride ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,3-Di(adamantan-1-yl)-1H-imidazol-3-ium chloride

07-0322-250MG
100202-856EA 162.77 USD
100202-856
1,3-Bis(1-adamantyl)imidazolium chloride ≥97%
1,3-Bis(1-adamantyl)imidazolium chloride

NHC

Formula: C₂₃H₃₃ClN₂
Melting Pt: 345…346 °C
MDL Number: MFCD03788919
CAS Number: 131042-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR