Zinpyr-1

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 9-(2-Carboxyphenyl)-2,7-dichloro-4,5-bis[di(2-pyridyl)aminomethyl]-6-hydroxy-3-xanthanone, 4′,5′-Bis[bis(2-pyridylmethyl)aminomethyl]-2′,7′-dichlorofluorescein

07-0314-10MG
100202-854EA 438.77 USD
100202-854
Zinpyr-1
Zinpyr-1
Formula: C₄₆H₃₆Cl₂N₆O₅
MDL Number: MFCD04039712
CAS Number: 288574-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR