Cesium iodide ≥99.9% (Cs basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cesiumiodide

93-5543-25G 93-5543-500G 93-5543-100G
100196-762EA 104.74 USD
100196-762 100199-286 100202-810
Cesium iodide ≥99.9% (Cs basis)
Cesium iodide
Formula: CsI
Boiling Pt: 1280 °C
Melting Pt: 621 °C
Density: 4.510
MDL Number: MFCD00010962
CAS Number: 7789-17-5
EINECS: 232-145-2
Merck Index: 12,02056

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR