Zinc oxide ≥99.999% (Zn basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Zinci oxidum, Zinc (II) oxide

93-3016-50G 93-3016-10G
100202-790EA 307.14 USD
100202-790 100209-090
Zinc oxide ≥99.999% (Zn basis), PURATREM
Zinc oxide
Formula: ZnO
Melting Pt: 1975 °C
Density: 5.606
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR