Bismuth ≥99.99%, pieces

Supplier: STREM CHEMICALS INC MS

83-0075-1000G 83-0075-250G
100202-780EA 307.14 USD
100202-780 100210-106
Bismuth ≥99.99%, pieces
Bismuth
Formula: Bi
Boiling Pt: 1560 °C
Melting Pt: 271.3 °C
Density: 9.80
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR