Osmium tetraoxide ≥99.95% (Os basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid
76-2950-5G
76-2950-500MG
76-2950-1G
76-2950-250MG
100204-648EA
833.66
USD
100204-648
100199-448
100202-688
100201-736
Osmium tetraoxide ≥99.95% (Os basis)
Osmium tetraoxide
Formula:
O₄Os Boiling Pt: 130 °C (1013 hPa) Melting Pt: 41…42 °C Density: 4.906 Storage Temperature: Ambient |
MDL Number:
MFCD00011150 CAS Number: 20816-12-0 EINECS: 244-058-7 UN: 2471 ADR: 6.1,I Merck Index: 13,06962 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...