Bis(ethylbenzene)chromium [mixture of (C2H5)xC6H6-x where x = 0-4)]

Supplier: STREM CHEMICALS INC MS
Danger

24-0145-5G 24-0145-25G 24-0145-1G
100208-390EA 232.12 USD
100208-390 100202-650 100209-788
Bis(ethylbenzene)chromium [mixture of (C2H5)xC6H6-x where x = 0-4)]
Bis(ethylbenzene)chromium

Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Boiling Pt: 140…160 °C (1 mmHg)
Density: 1,14…1.18
MDL Number: MFCD02091698
CAS Number: 12212-68-9
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR