Lanthanum ≥99.6%, chips

Supplier: STREM CHEMICALS INC MS
Danger

93-5746-100G 93-5746-25G
100207-114EA 553.42 USD
100207-114 100202-566
Lanthanum ≥99.6%, chips
Lanthanum
Formula: La
Boiling Pt: 3454 °C
Melting Pt: 920 °C
Density: 6.166
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR