Rubidium carbonate ≥99% (Rb basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-3703-50G
93-3703-10G
100202-554EA
300.06
USD
100202-554
100207-838
Rubidium carbonate ≥99% (Rb basis)
Rubidium carbonate
Formula:
CO₃Rb₂ Melting Pt: 837 °C Density: 2,43 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011188 CAS Number: 584-09-8 EINECS: 209-530-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...