Triethylborane ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-0540-100G
100202-522EA 622.77 USD
100202-522
Triethylborane ≥98%
Triethylborane

Metal Alkyls, Volatile Precursors for CVD

Formula: C₆H₁₅B
Boiling Pt: 95 °C
Melting Pt: –92,9 °C
Density: 0,6961 (23°)
Flash Pt: 32,8 °F
MDL Number: MFCD00009022
CAS Number: 97-94-9
UN: 2845
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR