N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

07-2050-500G 07-2050-100G
100202-356EA 359.51 USD
100202-356 100200-380
N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥99%
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Formula: C₆H₁₆N₂
Boiling Pt: 120…122 °C
Melting Pt: –55 °C
Density: 0.775
Flash Pt: 68 °F
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
EINECS: 203-744-6
UN: 1992
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR