Lead(IV) oxide ≥97% ACS
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-8210-100G
93-8210-500G
100202-316EA
100.49
USD
100202-316
100205-906
Lead(IV) oxide ≥97% ACS
Lead (IV) oxide
Formula:
O₂Pb Melting Pt: 290 °C (dec.) Density: 9.375 |
MDL Number:
MFCD00011165 CAS Number: 1309-60-0 EINECS: 215-174-5 UN: 1872 ADR: 5.1,III Merck Index: 12,05424 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...