Barium chloride, anhydrous ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-5652-500G 93-5652-100G
100202-210EA 161.35 USD
100202-210 100202-208
Barium chloride, anhydrous ≥98%
Barium chloride
Formula: BaCl₂
Boiling Pt: 1560 °C
Melting Pt: 963 °C
Density: 3.856
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 12,00998

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR