Tellurium dioxide ≥99.999% (Te basis), PURATREM

Supplier: STREM CHEMICALS INC MS

93-5216-50G 93-5216-10G
100202-194EA 345.35 USD
100202-194 100204-360
Tellurium dioxide ≥99.999% (Te basis), PURATREM
Tellurium dioxide
Formula: O₂Te
Boiling Pt: 450 °C (subl.)
Melting Pt: 733 °C
Density: 5.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,09123

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR