Niobium ≥99.8% Ta-0.1-1%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-4133-100G 93-4133-25G
100202-128EA 188.25 USD
100202-128 100197-856
Niobium ≥99.8% Ta-0.1-1%, powder
Niobium
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR