Copper(I) cyanide ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Copper cyanide, Cuprous cyanide
93-2916-500G
93-2916-100G
100202-050EA
181.17
USD
100202-050
100207-012
Copper(I) cyanide ≥99%
Copper (I) cyanide
Formula:
CCuN Boiling Pt: Dec. Melting Pt: 473 °C (in N₂) Density: 2.92 Storage Temperature: Ambient |
MDL Number:
MFCD00010975 CAS Number: 544-92-3 EINECS: 208-883-6 UN: 1587 ADR: 6.1,II Merck Index: 13,02691 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...