Iron(II) bromide hydrate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Iron dibromide hydrate

93-2628-100G 93-2628-25G
100202-024EA 428.86 USD
100202-024 100210-292
Iron(II) bromide hydrate
Iron (II) bromide hydrate
Formula: Br₂Fe·H₂O
Melting Pt: 27 °C
MDL Number: MFCD00016081
CAS Number: 13463-12-2
EINECS: 232-168-8
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR