Potassium tripolyphosphate ≥94%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassium triphosphate

93-1955-250G 93-1955-1000G
100197-726EA 36.8 USD
100197-726 100201-950
Potassium tripolyphosphate ≥94%
Potassium tripolyphosphate
Formula: K₅O₁₀P₃
Melting Pt: 620 °C
Density: 2,54
MDL Number: MFCD00054129
CAS Number: 13845-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR