Chlorotriethylsilane ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Triethylchlorosilane

93-1463-25G 93-1463-5G
100206-008EA 301.48 USD
100206-008 100201-918
Chlorotriethylsilane ≥99%
Chlorotriethylsilane
Formula: C₆H₁₅ClSi
Boiling Pt: 144 °C (735 mmHg)
Density: 0.8967 (20°)
Flash Pt: 85 °F
Storage Temperature: Ambient
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR