Aluminium iodide ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Aluminum iodide

93-1389-50G 93-1389-10G
100210-680EA 317.05 USD
100210-680 100201-904
Aluminium iodide ≥95%
Aluminium iodide
Formula: AlI₃
Boiling Pt: 360 °C
Melting Pt: 191 °C
Density: 3.98
Storage Temperature: Ambient
MDL Number: MFCD00003427
CAS Number: 7784-23-8
EINECS: 232-054-8
UN: 3260
ADR: 8,II
Merck Index: 12,00358

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR