Magnesium nitrate hexahydrate ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-1238-1000G
93-1238-250G
100206-924EA
111.82
USD
100206-924
100201-882
Magnesium nitrate hexahydrate ≥99% ACS
Magnesium nitrate hexahydrate
Formula:
Mg(NO₃)₂·6H₂O Boiling Pt: 330 °C (1013 hPa) Melting Pt: 89 °C Density: 1.64 Flash Pt: 93.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 EINECS: 233-826-7 UN: 1474 ADR: 5.1,III Merck Index: 13,05697 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...