Sodium hydrogensulfide, anhydrous

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Sodium hydrogen sulfide, Sodium hydrogen sulphide

93-1061-1G 93-1061-5G 93-1061-25G
100201-844EA 97.66 USD
100201-844 100201-848 100201-846
Sodium hydrogensulfide, anhydrous
Sodium hydrogensulphide
Formula: HNaS
Melting Pt: 350 °C
Density: 1.79 g/cm³ (20 °C)
MDL Number: MFCD00011124
CAS Number: 16721-80-5
EINECS: 240-778-0
UN: 2318
ADR: 4.2,II
Merck Index: 12,08730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR