Sodium perfluorooctanoate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Perfluorooctanoic acid, sodium salt

93-1010-25G 93-1010-5G
100201-836EA 410.46 USD
100201-836 100204-132
Sodium perfluorooctanoate
Sodium perfluorooctanoate
Formula: C₈F₁₅NaO₂
MDL Number: MFCD00040394
CAS Number: 335-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR