Rhenium ≥99.99%, powder, PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

75-1890-5G 75-1890-1G
100201-730EA 352.43 USD
100201-730 100201-728
Rhenium ≥99.99%, powder, PURATREM
Rhenium
Formula: Re
Boiling Pt: 5900 °C
Melting Pt: 3180 °C
Density: 21.04
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5
UN: 3089
ADR: 4.1,II
Merck Index: 12,08343

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR