Tetrakis(triphenylphosphine)palladium(0) ≥99% (99.9+%-Pd)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
46-2150-5G
46-2150-1G
100197-540EA
227.88
USD
100197-540
100201-684
Tetrakis(triphenylphosphine)palladium(0) ≥99% (99.9+%-Pd)
Tetrakis(triphenylphosphine)palladium (0)
Formula:
C₇₂H₆₀P₄Pd Melting Pt: 103…107 °C Density: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00010012 CAS Number: 14221-01-3 EINECS: 238-086-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...