Rhodium 5% on activated carbon wetted with 50-70% water, powder Evonik Noblyst® P3053

Supplier: STREM CHEMICALS INC MS
Warning

45-1863-10G
100201-668EA 856.31 USD
100201-668
Rhodium 5% on activated carbon wetted with 50-70% water, powder Evonik Noblyst® P3053
Rhodium
Formula: Rh
MW: 102.9 g/mol
MDL Number: MFCD00011201
CAS Number: 7440-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR