Zinc protoporphyrin
Supplier: STREM CHEMICALS INC MS
Synonyms:
Zinc(II) 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate, Protoporphyrin IX zinc(II), Zinc (II) Protoporphyrin IX
30-3090-10MG
30-3090-50MG
100196-196EA
67.94
USD
100196-196
100201-592
Zinc protoporphyrin
Zinc protoporphyrin
Formula:
C₃₄H₃₂N₄O₄Zn |
MDL Number:
MFCD00011612 CAS Number: 15442-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...