Chromium(III) acetylacetonate ≥97.5%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tris(2,4-pentanedionato-O,O')chromium (III), Chromium (III) 2,4-pentanedionate, Chromium (III) tris(2,4-pentanedionato-O,O')

24-0160-250G 24-0160-1000G 24-0160-50G
100203-050EA 152.86 USD
100203-050 100201-542 100205-014
Chromium(III) acetylacetonate ≥97.5%
Chromium (III) acetylacetonate

Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₅H₂₁CrO₆
Boiling Pt: 100 °C (0,2 mmHg) (subl.)
Melting Pt: 214 °C
Density: 1,266 g/cm³ (20 °C)
MDL Number: MFCD00000015
CAS Number: 21679-31-2
EINECS: 244-526-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR