2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate, 2-Acetoxyphenylboronic acid pinacol ester

05-1045-5G 05-1045-1G
100201-320EA 373.66 USD
100201-320 100201-318
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥97%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate

Boronic Acid and Ester

Formula: C₁₄H₁₉BO₄
Melting Pt: 46…48 °C
MDL Number: MFCD03453062
CAS Number: 480424-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR