(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R,R)-Et-DUPHOS, (-)-1,2-Bis((2R,5R)-2,5-diethylphospholano)benzene, 1,2-Bis((2R,5R)-2,5-diethylphospholan-1-yl)benzene

15-0097-2G 15-0097-100MG 15-0097-500MG
100201-180EA 1338.95 USD
100201-180 100208-814 100208-816
(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene ≥98%
(-)-1,2-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]benzene

BPE, DUPHOS, Phosphine

Formula: C₂₂H₃₆P₂
Density: 1,01
MDL Number: MFCD00142335
CAS Number: 136705-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR